Order Entry
Germany
ContactUsLinkComponent
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 96%
Toxic
Irritant
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 96%
Catalog # B24048.03
CAS Number:  
Toxic
Irritant
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 96%
Catalog # B24048.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

 :
 :259.151 g/mol
Melting Pt:147…152 °C
Storage Temperature:Refrigerator
EINECS:670-530-0
UN:2811
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:722-92-9
Formula:C₉H₇F₆NO
MDL Number:MFCD00039258