Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Amino-3-tert-butyl-1-methylpyrazole
5-Amino-3-tert-butyl-1-methylpyrazole 98%
5-Amino-3-tert-butyl-1-methylpyrazole 98%
5-Amino-3-tert-butyl-1-methylpyrazole 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
5-Amino-3-tert-butyl-1-methylpyrazole 98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
118430-73-2
:
Formula:
C₈H₁₅N₃
MDL Number:
MFCD00068002
:
153.22 g/mol
Melting Pt:
155…158 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB20095
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation