Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-tert-Butyl-2-methyl-3-furoic acid
5-tert-Butyl-2-methyl-3-furoic acid 97%
5-tert-Butyl-2-methyl-3-furoic acid 97%
5-tert-Butyl-2-methyl-3-furoic acid 97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
5-tert-Butyl-2-methyl-3-furoic acid 97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
38422-62-7
:
Formula:
C₁₀H₁₄O₃
MDL Number:
MFCD00221068
:
182.22 g/mol
Melting Pt:
90…94 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA19258
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation