Order Entry
Germany
ContactUsLinkComponent
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%
Catalog # A18181.06
CAS Number:  
Nɑ-Benzoyl-L-arginine ethyl ester hydrochloride ≥98%
Catalog # A18181.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

 :
 :342.82 g/mol
Melting Pt:127…131 °C
EINECS:220-157-0
CAS Number:2645-08-1
Formula:C₁₅H₂₂N₄O₃
MDL Number:MFCD00012579