N-tert-Butyl-ɑ-phenylnitrone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3376-24-7
:
Formula:C₁₁H₁₅NO
MDL Number:MFCD00008799
:177.25 g/mol
Merck Index:14,7056
Melting Pt:71…75 °C
Storage Temperature:Refrigerator
EINECS:222-168-6