Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Hydroxy-4-(trifluoromethyl)coumarin
7-Hydroxy-4-(trifluoromethyl)coumarin 98%
7-Hydroxy-4-(trifluoromethyl)coumarin 98%
Irritant
7-Hydroxy-4-(trifluoromethyl)coumarin 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
7-Hydroxy-4-(trifluoromethyl)coumarin 98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
575-03-1
:
Formula:
C₁₀H₅F₃O₃
MDL Number:
MFCD00037578
:
230.14 g/mol
Melting Pt:
178…180 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAA16537
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation