Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Strontium acetate hemihydrate
Strontium acetate hemihydrate 98%
Strontium acetate hemihydrate 98%
:
Thermo Fisher Scientific
Strontium acetate hemihydrate 98%
:
Thermo Fisher Scientific
:
14692-29-6
:
543-94-2
:
C₈H₁₄O₉Sr₂
:
MFCD00198064
:
14,8836
:
150 °C (-0.5H₂O)
:
429.43 g/mol
:
Ambient
:
208-854-8
msds_ALFAA16149
Thermo Scientific Acroseal 25 Years