

Sodium percarbonate 13 - 14% (active oxygen)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15630-89-4
:
Formula:2Na₂CO₃·3H₂O₂
MDL Number:MFCD00043204
:314.017 g/mol
EINECS:239-707-6
ADR:5.1,II
:GHS03
:GHS05
Storage Temperature:Ambient