Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3162-96-7
:
Formula:C₁₄H₁₈O₆
MDL Number:MFCD00006819
:282.29 g/mol
Melting Pt:163…166 °C
Storage Temperature:Refrigerator
EINECS:221-615-2