Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DL-Phenylalanine methyl ester hydrochloride
DL-Phenylalanine methyl ester hydrochloride 98%
DL-Phenylalanine methyl ester hydrochloride 98%
:
Thermo Fisher Scientific
DL-Phenylalanine methyl ester hydrochloride 98%
:
Thermo Fisher Scientific
:
5619-07-8
:
:
C₁₀H₁₄ClNO₂
:
MFCD00066113
:
215.677 g/mol
:
∼ 160 °C (dec.)
:
227-049-2
:
GHS07
:
Ambient
msds_ALFAA12758
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years