
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4644-61-5
:
Formula:C₈H₁₃NO₃
MDL Number:MFCD00043267
:207.65 g/mol
Melting Pt:∼ 172 °C (dec.)
EINECS:225-073-8
:GHS07
Storage Temperature:Ambient