Glove bags, small
Supplier: Thermo Fisher Scientific
Certificates
About this item
These polyethylene glove bags are ideal for laboratory use and field sampling.
Suitable for handling air and moisture sensitive, high purity chemicals and materials. Soluble in xylene.
Delivery: Supplied with closure, tubing coupler, and cotton gloves.