Oxone®, monopersulphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70693-62-8
:37222-66-5
Formula:2KHSO₅·KHSO₄·K₂SO₄
MDL Number:MFCD00040551
:615.50 g/mol
Density:1.15
Storage Temperature:Ambient
EINECS:274-778-7
UN:3084
ADR:8,II
:GHS03
:GHS08
:GHS05
:GHS07