Calcium pyrophosphate ≥96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-76-3
:
Formula:Ca₂O₇P₂
MDL Number:MFCD00015983
:254.097 g/mol
Melting Pt:1230 °C
Density:3.09
EINECS:232-221-5
:GHS07
Storage Temperature:Ambient