Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bis(di-tert-butylphenylphosphine)palladium(0)
Bis(di-tert-butylphenylphosphine)palladium(0) 98%
Bis(di-tert-butylphenylphosphine)palladium(0) 98%
Irritant
Bis(di-tert-butylphenylphosphine)palladium(0) 98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Bis(di-tert-butylphenylphosphine)palladium(0) 98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
52359-17-8
:
Formula:
C₂₈H₄₆P₂Pd
MDL Number:
MFCD15071400
:
551.04 g/mol
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA045853
Videos
Thermo Scientific Acroseal 25 Years
Documentation