0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
D-Biotin-N-succinimidyl ester
D-Biotin-N-succinimidyl ester ≥98%
D-Biotin-N-succinimidyl ester ≥98%
D-Biotin-N-succinimidyl ester ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
D-Biotin-N-succinimidyl ester ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
35013-72-0
:
Formula:
C₁₄H₁₉N₃O₅S
MDL Number:
MFCD00078531
:
341.38 g/mol
Melting Pt:
212…215 °C
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA044771
Videos
Thermo Scientific Acroseal 25 Years
Documentation