Trityl chloride resin 0.5 - 2.5 mmol/g on poly(styrene-divinylbenzene), cross-linked with 1% DVB 100-200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00284810
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05