Order Entry
Germany
ContactUsLinkComponent
Nickel plating solution, electroless ammonia-free
Health
Irritant
Nickel plating solution, electroless ammonia-free
Catalog # 44070.AK
CAS Number:  
Health
Irritant
Nickel plating solution, electroless ammonia-free
Catalog # 44070.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3082
ADR:9,III
 :GHS08
 :GHS07
MDL Number:MFCD00011137