Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Fluorescamine

Fluorescamine
Certificates
Fluorescamine

3 Options Available Below

Product Details & Documents

CAS No.: 38183-12-9 Molecular Formula: C17H10O4 Formula Weight: 278.27 Physical Form: Powder Appearance: White to cream Odour: Odourless Melting Point: 155-158° MDL No.: MFCD00005928

Applications: For fluorescent detection of amino acids, peptides and proteins

Synonyms:  Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
  • CAS Number:38183-12-9
  • Formula:C₁₇H₁₀O₄
  • MDL Number:MFCD00005928
  • Molecular Weight:278.26 g/mol
  • Merck Index:14,4158
  • Melting Pt:155…158 °C
  • Storage Temperature:Ambient
  • EINECS:253-814-5
Product Family Options

Product Information