Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Formaldehyde
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
:
Thermo Fisher Scientific
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
:
Thermo Fisher Scientific
:
50-00-0
:
:
CH₂O
:
MFCD00003274
:
30.03 g/mol
:
3334
:
9,III
:
GHS08
:
GHS07
:
Ambient
Concentration (w/v):
15.5 to 16.5%
Appearance:
Clear colourless liquid
msds_ALFA043368
Thermo Scientific Acroseal 25 Years
Thermo Scientific Paraformaldehyde