Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium
Aluminium ≥99% (metals basis), spheres, Ø 6.35 mm (0.25 in)
Aluminium ≥99% (metals basis), spheres, Ø 6.35 mm (0.25 in)
Aluminium ≥99% (metals basis), spheres, Ø 6.35 mm (0.25 in)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Aluminium ≥99% (metals basis), spheres, Ø 6.35 mm (0.25 in)
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
7429-90-5
:
Formula:
Al
MDL Number:
MFCD00134029
:
26.98 g/mol
Storage Temperature:
Ambient
EINECS:
231-072-3
Related Information
Documentation
SDS
msds_ALFA042030
Documentation