Order Entry
Germany
ContactUsLinkComponent
Sulfur in crude oil standard solutions, Specpure®
Sulfur in crude oil standard solutions, Specpure®
Certificates
Sulfur in crude oil standard solutions, Specpure®
Certificates

Specifications

  • MDL No.:
    MFCD00197838
  • UN No.:
    1267
  • Flash point:
    25°C(77°F)
  • Physical form:
    Liquid
  • Odour/scent:
    Unpleasant
  • Appearance:
    Black

Specifications