Order Entry
Germany
ContactUsLinkComponent
 
Sulfur in isooctane standard solutions, Specpure®
 
Sulfur in isooctane standard solutions, Specpure®
 

 

  • MDL No.:
    MFCD00804269
  • UN No.:
    1262
  • Physical form:
    Liquid
  • Odour/scent:
    Petrol-like
  • Appearance:
    Colourless