Polyvinyl alcohol, M.W. ∼ 57,000-66,000 98-99%, medium molecular weight, hydrolysed
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9002-89-5
:25213-24-5
Formula:[-CH₂CH(OH)-]n
MDL Number:MFCD00081922
Storage Temperature:Ambient