Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stainless steel
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.51 mm (0.02 in) Type 304
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.51 mm (0.02 in) Type 304
:
Thermo Fisher Scientific
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.51 mm (0.02 in) Type 304
:
Thermo Fisher Scientific
:
:
Fe
:
MFCD00148499
:
55.845 g/mol
:
GHS08
:
GHS07
:
Ambient
msds_ALFA040946