Carbon black ≥99.9%, acetylene, 50% compressed
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1333-86-4
:
Formula:C
MDL Number:MFCD00133992
:12.01 g/mol
Storage Temperature:Ambient
EINECS:215-609-9
UN:1325
ADR:4.1,III
Specification Test Results
- Trace Metal:99.9+%