Order Entry
Germany
ContactUsLinkComponent
 
Sulfur in diesel fuel standard solutions, Specpure®
 
Sulfur in diesel fuel standard solutions, Specpure®
 

 

  • MDL No.:
    MFCD01095233
  • Physical form:
    Liquid
  • Odour/scent:
    Petroleum-like
  • Appearance:
    Yellow