Single crystal substrates
A single crystal is a material in which the crystal lattice of the entire sample is continuous and unbroken to the edges of the sample, with no grain boundaries.
The absence of the defects associated with grain boundaries can give single crystals unique properties, particularly mechanical, optical and electrical. These properties, in addition to making them precious in some gems, are industrially used in technological applications, especially in optics and electronics.
A single crystal is a material in which the crystal lattice of the entire sample is continuous and unbroken to the edges of the sample, with no grain boundaries.
The absence of the defects associated with grain boundaries can give single crystals unique properties, particularly mechanical, optical and electrical. These properties, in addition to making them precious in some gems, are industrially used in technological applications, especially in optics and electronics.
Documents
- Catalog No:
- 38509.KT
- 38509.KU
- MDL No.:
- MFCD01868885
- MFCD01868885
- Molecular formula:
- ZrO2 + 9.5% Y2O3
- ZrO2 + 9.5% Y2O3
Product Family Options
- Description
- Zirconium oxide/yttrium oxide substrate, 10×10×1 mm, polished one side, 100 orientation1 * 1 ST108,00 €
Specifications
Cat. No.38509.KTMDL No.MFCD01868885Molecular formulaZrO2 + 9.5% Y2O3 - Zirconium oxide/yttrium oxide substrate, 10×10×1 mm, polished one side, 100 orientation1 * 5 ST434,00 €
Specifications
Cat. No.38509.KUMDL No.MFCD01868885Molecular formulaZrO2 + 9.5% Y2O3
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



