Order Entry
Germany
ContactUsLinkComponent
Palladium 5% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Palladium 5% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Artikelnr. 38301.22
CAS Number:  
Palladium 5% on activated carbon powder wetted with ca. 50% water for use as a catalyst, unreduced
Artikelnr. 38301.22
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :106.420 g/mol
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167