

1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38465-86-0
:
Formula:C₃₄H₃₈F₆IrP₃
MDL Number:MFCD00064800
:845.79 g/mol
Melting Pt:224 °C (dec.)
:GHS06
:GHS07
Storage Temperature:Ambient