VWR® Rome, Safety Spectacles
111-1842
- Type:Anti-UV (2-1,2), anti-scratch, anti-fog
- Colour:Blue
- Lenses:Clear PC
- Pk:1
Wrap-around protective eyewear offering a comfortable and custom fit all day long.
- Adjustable temple length enables wearers to fit the eyewear to their facial profile
- Lightweight, frameless design
- Soft nose bridge for added comfort
- Easy lens replacement
: Compliant with EN 166, EN 170; EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)