VWR® Montreal, Safety Goggles
Catalog # 111-1839
Supplier: VWR Collection
Specifications
- Type:Anti-scratch, anti-fog
- Colour:Black
- Pk:1
Specifications
About this item
Ergonomically designed wide vision goggles with co-injected frame provides 180° panoramic view. PC lens, PP (hard part) and AES Santoprene (soft part) frame, polyester-rubber headband.
- Indirect ventilation to prevent fogging
- Withstands extreme temperatures
- Designed to give optimum fit to all facial contours
- Easy four-step lens replacement
Certifications: Compliant with: EN 166:2001. Frame: EN 166 3 9 BT CE. Lens: VWR® 1 BT 9.