Orders LinkContactUsLinkComponent
Test kits, silicate, Spectroquant®, Supelco®
Test kits, silicate, Spectroquant®, Supelco®
Test kits, silicate, Spectroquant®, Supelco®
Supplier:  Merck
Certificates
Test kits, silicate, Spectroquant®, Supelco®
Test kits, silicate, Spectroquant®, Supelco®
3 Options Available Below
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60 A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.01813.0001
  • 1.14794.0001
  • 1.00857.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Range
    Chemical method
    For unit
    Pk
    Availability
    Price
  • 1.01813.0001
    Silicate test
    Silicate test
    0,0005 - 0,500 mg/L SiO₂
    Silicomolybdenum blue
    Prove, Nova 60, Multy, Move
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 162,00 €
    162,00 €
    1 * 100 Tests
       162,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.01813.0001
    M-Clarity quality level
  • 1.14794.0001
    Silicate (silicic acid) test
    Silicate (silicic acid) test
    0,005 - 5 mg/L Si , 00,11 - 10,7 mg/L SiO₂
    Silicomolybdenum blue
    Prove, Nova 60, Multy, Move
    300 Tests
    Log in for availability
    Log in for availability
    1 * 300 Tests
    1 * 300 Tests - 210,00 €
    210,00 €
    1 * 300 Tests
       210,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14794.0001
    M-Clarity quality level
    MQ100
  • 1.00857.0001
    Silicate (silicic acid) test
    Silicate (silicic acid) test
    0,5 - 500 mg/L Si, 1,1 - 1070 mg/L SiO₂
    Molybdosilicate
    Prove, Nova 60, Multy, Move
    100 Tests
    Log in for availability
    Log in for availability
    1 * 100 Tests
    1 * 100 Tests - 177,00 €
    177,00 €
    1 * 100 Tests
       177,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.00857.0001
    M-Clarity quality level
    MQ100