Auftragserfassung
Germany
ContactUsLinkComponent
Testsätze, Molybdän, Spectroquant®, Supelco®
  1.00860.0001
 :  Merck
 :  
undefined
Testsätze, Molybdän, Spectroquant®, Supelco®
  1.00860.0001
 :  Merck
 :  
  • Chemische Methode:
    Bromopyrogallolrot
  • VE:
    25 Tests
  • Bezeichnung:
    Molybdän-Küvettentest
  • M-Clarity Qualitätsstufe:
    MQ100
  • Für Gerät:
    Prove, Nova 30/60, Multy
  • Messbereich:
    0,02 - 1,00 mg/l Mo
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits.
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.