Order Entry
Germany
ContactUsLinkComponent
 
Pure water system, PURELAB® Chorus 2 (RO/DI)
  ELGAPC2RODIM1
undefined
Pure water system, PURELAB® Chorus 2 (RO/DI)
  ELGAPC2RODIM1

 

  • Flow rate:
    0,33 l/min
  • Description:
    Water purification system, PURELAB® Chorus 2, 20 l/hr
  • Model:
    PURELAB® Chorus 2
  • Bacteria:
    <5 CFU/ml
  • Average daily needs:
    up to 480 l/day
  • Conductivity:
    <2000 µS/cm
  • Feed water:
    Potable mains water supply
  • Max. operating pressure:
    2 bar
  • Particles:
    >99 % rejection
  • TOC content:
    <30 ppb
  • Weight:
    18 kg
  • W×D×H:
    375×340×435 mm
  • Pk:
    1

 

 

This flexible system reliably delivers Type II water.

  • Reverse Osmosis feed contains optimised resin mixes to maximise consumables capacity
  • Easy to install, operate and maintain with a clear indication of water purity
  • Optional CO2 removal from the purified water (post RO) increases the life of downstream consumables
  • Option to reduce water consumption for low hardness feed waters
  • Modular: Multiple PURELAB Chorus 2 units can feed into one reservoir and systems can be expanded post-installation

Typical applications: Stills replacement, buffer preparation, pH solution preparation, washing / rinsing, autoclaves, general chemistry, hydroponics, steam generators, steriliser feed, feed to Type I polishers.

The system can be combined with a range of storage reservoirs (15, 30, 60 or 100 l - not included) designed to maintain the optimum purity of stored purified water.