ContactUsLinkComponent
Industrial gloves, uvex phynomic C3
 :  UVEX
undefined
Industrial gloves, uvex phynomic C3
 :  UVEX
  • Catalog No:
  • 112-4947
  • 112-4945
  • 112-4946
  • 112-4944
  • 112-4943
  • 112-4942
  • Model:
  • C3
  • C3
  • C3
  • C3
  • C3
  • C3
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide/elastane
  • Polyamide/elastane
  • Polyamide/elastane
  • Polyamide/elastane
  • Polyamide/elastane
  • Polyamide/elastane
  • Finish:
  • Aqua-polymer foam coating on palm and fingertips
  • Aqua-polymer foam coating on palm and fingertips
  • Aqua-polymer foam coating on palm and fingertips
  • Aqua-polymer foam coating on palm and fingertips
  • Aqua-polymer foam coating on palm and fingertips
  • Aqua-polymer foam coating on palm and fingertips
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X43B)
  • EN 388:2016 (4X43B)
  • EN 388:2016 (4X43B)
  • EN 388:2016 (4X43B)
  • EN 388:2016 (4X43B)
  • EN 388:2016 (4X43B)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
  • +