Surface active cleaning agent, Neutrad®
Catalog # DECNNEUT5
Supplier: DECON LAB
Specifications
- Capacity:5 l
- Description:Surface active cleaning agent, Neutrad®
- Pk:5 L
- pH value:7
Specifications
About this item
A surface active cleaning agent and/or decontaminant, for use on materials which may be corroded, etched, discoloured or weakened by acidic or alkaline cleaning agents. Ideal for cleaning all non ferrous metals such as aluminium, zinc, copper, silver and brass.
- Phosphate- and enzyme-free
- Non toxic
- Neutral pH 7
OECD 301E (ISO method 7827-1986 (E))