Aluminium headspace caps
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.
- Temperature resistance up to 130 °C
- Very economical and has low resistance to coring
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.