Orders LinkContactUsLinkComponent
GC guard columns, HydroGOLD
GC guard columns, HydroGOLD
GC guard columns, HydroGOLD
Certificates
GC guard columns, HydroGOLD
7 Options Available Below
Product Details & Documents
These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.
  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • Protects against column contamination caused by non-volatile materials
Product Family Options
Product Information
  • Column Format
    Ø int.
    Length
    Availability
    Price
  • Guard column
    0,32 mm
    2 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 78,70 €
    78,70 €
    1 * 1 ST
       78,70 €
  • Guard column
    0,53 mm
    2 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,20 €
    86,20 €
    1 * 1 ST
       86,20 €
  • Guard column
    0,25 mm
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 103,00 €
    103,00 €
    1 * 1 ST
       103,00 €
  • Guard column
    0,32 mm
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • Guard column
    0,53 mm
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 139,00 €
    139,00 €
    1 * 1 ST
       139,00 €
  • Guard column
    0,32 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €
  • Guard column
    0,53 mm
    10 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 288,00 €
    288,00 €
    1 * 1 ST
       288,00 €