Safety ankle boots, Velcro® fastening, U-Special, Bulls
Supplier: U-Power
Certificates
About this item
Safety ankle boots made from water resistant leather and a heat resistant to 300 °C, antislip nitrile rubber sole.
- Comfortable, anatomically shaped ergo dry insole features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
- Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
- Stress-out tongue system allows easy instep and provides added comfort when walking
- Natural fit width 11 for additional comfort
- Air toe protective aluminium toecap with breathable membrane (200 J)
- Puncture resistant steel midsole
Colour: Black
Colour: Black
Certifications: EN 20345:2011; S3, SRC, HI, HRO