ContactUsLinkComponent
Safety ankle boots, lace-up, Step One, Pitucon
Safety ankle boots, lace-up, Step One, Pitucon
Catalog # UPOWSO10043/42
Supplier:  U-Power
undefined
Safety ankle boots, lace-up, Step One, Pitucon
Catalog # UPOWSO10043/42
Supplier:  U-Power
Specifications
  • Shoe size:
    42
  • Colour:
    Black
  • Pk:
    1 PAIR
Specifications
About this item
Safety ankle boots made from water resistant leather plus a slip resistant, dual-density PU sole.
  • Ergo Dry comfortable, anatomically-shaped insole features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • WingTex tunnel lining, with honeycomb structure, ensures breathability and excellent absorbent features
  • Stress-out tongue system allows easy instep providing added comfort when walking
  • Natural fit (width 11) for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Puncture resistant steel midsole
Colour: Black
Colour: Black
Certifications: EN 20345:2011; S3, SRC.