ContactUsLinkComponent
Safety tube holders, VACUETTE® QUICKSHIELD
Catalog # GREI450230_1000
Supplier:  Greiner Bio-One
undefined
Safety tube holders, VACUETTE® QUICKSHIELD
Catalog # GREI450230_1000
Supplier:  Greiner Bio-One
Specifications
  • Description:
    Plastic, standard
  • Seq:
    01
  • Pk:
    1.000
Specifications
About this item
The VACUETTE® QUICKSHIELD safety tube holder is especially suitable for routine blood collection.
  • Safety shield activated with the aid of a solid surface and/or thumb
  • Comfort through one-handed activation
  • User-friendly, dot on needle container allows user to choose which side
  • Especially safe as the fingers always remain behind the needle tip
  • The protective cap is pre-attached to the holder, remaining stable; once activated there is no chance of the protective cap coming off
Exclusively for use with VACUETTE® multiple use drawing needles or VACUETTE® VISIO PLUS needles.
As combination product with a pre-mounted VACUETTE® VISIO PLUS needle, the VACUETTE® QUICKSHIELD Complete PLUS enables immediate visual control of blood flow to confirm successful venipuncture.