Order Entry
Germany
ContactUsLinkComponent
1-Butanesulphonic acid sodium salt ≥98%, J.T.Baker®
1-Butanesulphonic acid sodium salt ≥98%, J.T.Baker®
Catalog # D600-03
Supplier:  Avantor
CAS Number:  
undefined
1-Butanesulphonic acid sodium salt ≥98%, J.T.Baker®
Catalog # D600-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 98 %
  • Assay (C₄H₉NaO₃) (by titration):
    ≥98 %
  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
 :160.17 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:219-201-1
CAS Number:2386-54-1
Formula:C₄H₉NaO₃S
MDL Number:MFCD00007540