Trichloroacetic acid (TCA) ≥99.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0414-04
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth, black T/E cap
- Pk:125 g
- Seq:3445
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:Passes Test
- IMPORTANT: Store or transport at or below 30°C (86°F).:≤0.01%
- Assay (Cl₃CCOOH) (by acidimetry):≥99.0 %
- Clarity of Solution:Passes Test
- Insoluble Matter:≤0.01 %
- Residue after Ignition:≤0.03 %
- Chloride (Cl):≤0.002 %
- Nitrate (NO₃):≤0.002 %
- ACS - Phosphate (PO₄):≤5 ppm
- Sulfate (SO₄):≤0.02 %
- Heavy Metals (as Pb):≤0.002 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Substances Darkened by H₂SO₄:Passes Test
Specification Test Results
About this item
:
:163.39 g/mol
Merck Index:13,09700
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.63 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
EINECS:200-927-2
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:76-03-9
Formula:Cl₃CCOOH
MDL Number:MFCD00004177