Safety shoes, lace-up, Entry, Simple
Supplier: U-Power
Certificates
About this item
Safety shoes made from water resistant leather with a PU scuff cap and a slip resistant, hard-wearing PU sole.
- Breathable 3D lining
- Removable, anatomically shaped insole
- Quick release lacing system
- Natural fit width 11 for additional comfort
- Protective steel toecap (200 J)
- Puncture resistant steel midsole
Colour: Black
Certifications: EN 20345:2011; S1P, SRC