uvex unidur 6649 Cut Resistant Gloves
: UVEX
These mottled grey HPPE/PA/elastane gloves with PU coating are ideal for both clean and dirty environments. The HPPE fibre ensures good cut protection (level 3).
- Good cut resistance
- High abrasion resistance, high flexibility
- Highest level of durability – ideal for challenging environments
- Outstanding dexterity
Ideal for use in the automotive and metal industry as well as for packaging. Suitable for dry and slightly moist areas.
: EN 388:2016 (4342B).
- Catalog No:
- 112-4502
- 112-4500
- 112-4499
- 112-4503
- 112-4501
- Model:
- Unidur 6649
- Unidur 6649
- Unidur 6649
- Unidur 6649
- Unidur 6649
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Finish:
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- PU coating on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)