Industrial gloves, uvex phynomic XG
: UVEX
New Product
The uvex phynomic XG offers the best oil grip in its class. The polyamide/elastane liners feature an innovative aqua-polymer Xtra Grip foam coating which is also particularly flexible and extremely robust.
- Ultra-slow liquid breakthrough provides additional skin protection
- High abrasion resistance, high levels of dexterity
- Second skin fit gives a natural touch
- Flexible construction reduces fatigue
- Free from allergenic substances - no harmful substances used in the manufacturing
- Suitable for the most allergy-sensitive wearers
Ideal for fine assembly work/assembly work, general maintenance work and precision work. Suitable for use in damp and very oily conditions.
: EN 388:2016 (4121X)
OEKO-Tex® Standard 100
OEKO-Tex® Standard 100
- Catalog No:
- 112-4523
- 112-4427
- 112-4423
- 112-4426
- 112-4424
- UVEX6007005
- UVEX6007012
- 112-4425
- Model:
- XG
- XG
- XG
- XG
- XG
- XG
- XG
- XG
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Polyamide/elastane
- Finish:
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Aqua polymer Xtra Grip foam coating on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)