Nickel wetted with ca. 50% water catalyst (Raney®-type), for synthesis, Sigma-Aldrich®
: Merck
:7440-02-0
:
:Ni
:MFCD00011137
:58.69 g/mol
:4.2,II
:M-Clarity™ quality level = MQ200
:1378
:GHS02
:GHS08
:GHS07
:Ambient
- Assay of Ni, calc. on dried substance:≥ 90.0 %
- Assay od Al, calc. on dried substance:≤ 10.0 %
- Suitable for hydrogenation catalyst:passes test