ContactUsLinkComponent
Affinity chromatography columns, tagged package, His
Affinity chromatography columns, tagged package, His
Catalog # 29-0588-03
Supplier:  Cytiva
undefined
Affinity chromatography columns, tagged package, His
Catalog # 29-0588-03
Supplier:  Cytiva
Specifications
  • Description:
    Tagged package, His
  • Pk:
    1 KIT
Specifications
About this item
His package is designed for the convenient purification of histidine-tagged proteins.
The His package comes with ready-to-use columns packed with high performance media developed for high yield (HisTrap™ HP, 1×1 ml) or high purity (HiTrap™ TALON crude, 1×1 ml) and a HiTrap™ desalting column (1×5 ml) for removing imidazole from eluate.
Delivery: The kit contains one of each HisTrap™ HP (1×1 ml), HiTrap™ TALON crude (1×1 ml) and HiTrap™ Desalting (1×5 ml) columns.