Order Entry
Germany
ContactUsLinkComponent
Sepharose™ CL-2B Gel Filtration Base Matrix
Sepharose™ CL-2B Gel Filtration Base Matrix
Catalog # 17-0140-01
Supplier:  Cytiva
CAS Number:  
Sepharose™ CL-2B Gel Filtration Base Matrix
Catalog # 17-0140-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    60 - 200 µm
  • Pk:
    1 L
  • Description:
    Sepharose™ CL-2B
  • pH range:
    3 - 13
  • Matrix:
    2% agarose
  • Storage temperature:
    4 - 30 °C
  • Description:
    Sepharose™ CL-2B
  • Flow rate:
    <15 cm/h
  • Exclusion limit:
    >4 × 107
  • Type:
    Liquid

Specifications

About this item

Sepharose CL-2B is a well-proven cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.

  • 2% cross-linked agarose gel filtration media
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability